4-Ethoxy-2-methyl-phenol 95%

Lieferant: Apollo Scientific

OR957503-1G
APOSOR957503-1GEA 632 EUR
APOSOR957503-1G
4-Ethoxy-2-methyl-phenol 95%
4-Ethoxy-2-methyl-phenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24715102
CAS-Nummer: 84822-50-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR