3-Ethoxy-5-methylphenol 95%

Lieferant: Apollo Scientific

OR957504-1G
APOSOR957504-1GEA 507 EUR
APOSOR957504-1G
3-Ethoxy-5-methylphenol 95%
3-Ethoxy-5-methylphenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24714517
CAS-Nummer: 24741-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR