2-(2-Ethoxyethoxy)phenol 96%

Lieferant: Apollo Scientific

OR957505-1G
APOSOR957505-1GEA 0 EUR
APOSOR957505-1G
2-(2-Ethoxyethoxy)phenol 96%
2-(2-Ethoxyethoxy)phenol
Formel: C₁₀H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12174696
CAS-Nummer: 62621-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR