4-Methoxy-3,5-dimethylphenol 95%
Lieferant: Apollo Scientific
OR957506-1G
APOSOR957506-1GEA
212
EUR
APOSOR957506-1G
4-Methoxy-3,5-dimethylphenol 95%
4-Methoxy-3,5-dimethylphenol
Formel:
C₉H₁₂O₂ Molecular Weight: 152,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18432122 CAS-Nummer: 4962-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...