2-Chloro-5-methoxypyridin-4-ol 96%
Lieferant: Apollo Scientific
|
Achtung
|
OR957507-1G
APOSOR957507-1GEA
203
EUR
APOSOR957507-1G
2-Chloro-5-methoxypyridin-4-ol 96%
2-Chloro-5-methoxypyridin-4-ol
Formel:
C₆H₆ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189881 CAS-Nummer: 1196145-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...