3-Chloro-5-(hydroxymethyl)phenol 95%
Lieferant: Apollo Scientific
OR957512-1G
APOSOR957512-1GEA
632
EUR
APOSOR957512-1G
3-Chloro-5-(hydroxymethyl)phenol 95%
3-Chloro-5-(hydroxymethyl)phenol
Formel:
C₇H₇ClO₂ Molecular Weight: 158,582 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234658 CAS-Nummer: 885270-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...