4-[(Dimethylamino)methyl]phenol 95%

Lieferant: Apollo Scientific

OR957515-1G
APOSOR957515-1GEA 1250 EUR
APOSOR957515-1G
4-[(Dimethylamino)methyl]phenol 95%
4-[(Dimethylamino)methyl]phenol
Formel: C₉H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00053408
CAS-Nummer: 103-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR