3-[(Dimethylamino)methyl]phenol 95%

Lieferant: Apollo Scientific

Synonyme: 3-((Dimethylamino)methyl)phenol

OR957516-1G
APOSOR957516-1GEA 165 EUR
APOSOR957516-1G
3-[(Dimethylamino)methyl]phenol 95%
3-[(Dimethylamino)methyl]phenol
Formel: C₉H₁₃NO
Molecular Weight: 151,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00081105
CAS-Nummer: 60760-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR