2,6-Dichloro-3-methylphenol 95%

Lieferant: Apollo Scientific

OR957526-1G
APOSOR957526-1GEA 632 EUR
APOSOR957526-1G
2,6-Dichloro-3-methylphenol 95%
2,6-Dichloro-3-methylphenol
Formel: C₇H₆Cl₂O
Molecular Weight: 177.028 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061909
CAS-Nummer: 13481-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR