3-Hydroxy-4-propoxy-benzonitrile 95%

Lieferant: Apollo Scientific

OR957527-1G
APOSOR957527-1GEA 632 EUR
APOSOR957527-1G
3-Hydroxy-4-propoxy-benzonitrile 95%
3-Hydroxy-4-propoxy-benzonitrile
Formel: C₁₀H₁₁NO₂
Molecular Weight: 177,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18826309
CAS-Nummer: 518058-74-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR