3-(2-Methylpropoxy)phenol 95%
Lieferant: Apollo Scientific
OR957529-1G
APOSOR957529-1GEA
0
EUR
APOSOR957529-1G
3-(2-Methylpropoxy)phenol 95%
3-(2-Methylpropoxy)phenol
Formel:
C₁₀H₁₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11181934 CAS-Nummer: 91950-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...