4-(2-Methylpropoxy)phenol 95%

Lieferant: Apollo Scientific

OR957530-1G
APOSOR957530-1GEA 538 EUR
APOSOR957530-1G
4-(2-Methylpropoxy)phenol 95%
4-(2-Methylpropoxy)phenol
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11181741
CAS-Nummer: 56069-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR