4-Isopropoxy-3-methylphenol 95%

Lieferant: Apollo Scientific

OR957533-1G
APOSOR957533-1GEA 632 EUR
APOSOR957533-1G
4-Isopropoxy-3-methylphenol 95%
4-Isopropoxy-3-methylphenol
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14535469
CAS-Nummer: 1216201-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR