4-Ethoxy-3,5-dimethylphenol 95%

Lieferant: Apollo Scientific

OR957536-1G
APOSOR957536-1GEA 632 EUR
APOSOR957536-1G
4-Ethoxy-3,5-dimethylphenol 95%
4-Ethoxy-3,5-dimethylphenol
Formel: C₁₀H₁₄O₂
Molecular Weight: 166,217 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24726029
CAS-Nummer: 217448-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR