5-Chloro-2-ethoxyphenol 95%
Lieferant: Apollo Scientific
OR957538-1G
APOSOR957538-1GEA
632
EUR
APOSOR957538-1G
5-Chloro-2-ethoxyphenol 95%
5-Chloro-2-ethoxyphenol
Formel:
C₈H₉ClO₂ Molecular Weight: 172,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14534966 CAS-Nummer: 57428-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...