4-Chloro-2-ethoxyphenol 95%

Lieferant: Apollo Scientific

OR957540-1G
APOSOR957540-1GEA 632 EUR
APOSOR957540-1G
4-Chloro-2-ethoxyphenol 95%
4-Chloro-2-ethoxyphenol
Formel: C₈H₉ClO₂
Molecular Weight: 172,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24714153
CAS-Nummer: 18113-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR