3-Chloro-4-ethoxy-phenol 95%

Lieferant: Apollo Scientific

OR957541-1G
APOSOR957541-1GEA 632 EUR
APOSOR957541-1G
3-Chloro-4-ethoxy-phenol 95%
3-Chloro-4-ethoxy-phenol
Formel: C₈H₉ClO₂
Molecular Weight: 172,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14536966
CAS-Nummer: 1216134-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR