4-Chloro-3-ethoxyphenol 95%

Lieferant: Apollo Scientific

OR957542-1G
APOSOR957542-1GEA 632 EUR
APOSOR957542-1G
4-Chloro-3-ethoxyphenol 95%
4-Chloro-3-ethoxyphenol
Formel: C₈H₉ClO₂
Molecular Weight: 172,609 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440337
CAS-Nummer: 685535-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR