3-Ethoxy-4-methoxyphenol 95%

Lieferant: Apollo Scientific

OR957545-1G
APOSOR957545-1GEA 0 EUR
APOSOR957545-1G
3-Ethoxy-4-methoxyphenol 95%
3-Ethoxy-4-methoxyphenol
Formel: C₉H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00009093
CAS-Nummer: 65383-57-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR