2-Chloro-5-ethoxyphenol 95%
Lieferant: Apollo Scientific
OR957547-1G
APOSOR957547-1GEA
632
EUR
APOSOR957547-1G
2-Chloro-5-ethoxyphenol 95%
2-Chloro-5-ethoxyphenol
Formel:
C₈H₉ClO₂ Molecular Weight: 172,609 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24721838 CAS-Nummer: 89981-62-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...