2-[(Cyclopropylamino)methyl]phenol 95%

Lieferant: Apollo Scientific

OR957553-1G
APOSOR957553-1GEA 364 EUR
APOSOR957553-1G
2-[(Cyclopropylamino)methyl]phenol 95%
2-[(Cyclopropylamino)methyl]phenol
Formel: C₁₀H₁₃NO
Molecular Weight: 163,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09047227
CAS-Nummer: 643007-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR