4-Cyclopentylphenol 95%

Lieferant: Apollo Scientific

OR957556-1G
APOSOR957556-1GEA 183 EUR
APOSOR957556-1G
4-Cyclopentylphenol 95%
4-Cyclopentylphenol
Formel: C₁₁H₁₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD00009953
CAS-Nummer: 1518-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR