4-(Cyclopropylmethoxy)phenol 95%

Lieferant: Apollo Scientific

OR957557-1G
APOSOR957557-1GEA 806 EUR
APOSOR957557-1G
4-(Cyclopropylmethoxy)phenol 95%
4-(Cyclopropylmethoxy)phenol
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12138943
CAS-Nummer: 63659-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR