3-Hydroxybenzoyl chloride 95%

Lieferant: Apollo Scientific

OR957559-1G
APOSOR957559-1GEA 364 EUR
APOSOR957559-1G
3-Hydroxybenzoyl chloride 95%
3-Hydroxybenzoyl chloride
Formel: C₇H₅ClO₂
MDL: MFCD11169648
CAS-Nummer: 40812-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR