3-(Cyclopropylmethoxy)phenol 95%

Lieferant: Apollo Scientific

OR957560-1G
APOSOR957560-1GEA 632 EUR
APOSOR957560-1G
3-(Cyclopropylmethoxy)phenol 95%
3-(Cyclopropylmethoxy)phenol
Formel: C₁₀H₁₂O₂
Molecular Weight: 164,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12138944
CAS-Nummer: 1154342-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR