2-(Cyclohexyloxy)pyridin-3-ol 95%

Lieferant: Apollo Scientific

OR957563-1G
APOSOR957563-1GEA 632 EUR
APOSOR957563-1G
2-(Cyclohexyloxy)pyridin-3-ol 95%
2-(Cyclohexyloxy)pyridin-3-ol
Formel: C₁₁H₁₅NO₂
Molecular Weight: 193,242 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17000261
CAS-Nummer: 1243284-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR