4-Methoxy-2,6-dimethylphenol 96%

Lieferant: Apollo Scientific

OR957565-1G
APOSOR957565-1GEA 237 EUR
APOSOR957565-1G
4-Methoxy-2,6-dimethylphenol 96%
4-Methoxy-2,6-dimethylphenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14537195
CAS-Nummer: 2431-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR