2'-Chloro-5'-hydroxyacetophenone 95%
Lieferant: Apollo Scientific
Synonyme:
1-(2-Chloro-5-hydroxyphenyl)ethanone
OR957566-1G
APOSOR957566-1GEA
248
EUR
APOSOR957566-1G
2'-Chloro-5'-hydroxyacetophenone 95%
2'-Chloro-5'-hydroxyacetophenone
Formel:
C₈H₇ClO₂ Molecular Weight: 170,593 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393141 CAS-Nummer: 58020-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...