3-Chloro-4-isopropoxyphenol 96%

Lieferant: Apollo Scientific

OR957570-1G
APOSOR957570-1GEA 364 EUR
APOSOR957570-1G
3-Chloro-4-isopropoxyphenol 96%
3-Chloro-4-isopropoxyphenol
Formel: C₉H₁₁ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14537050
CAS-Nummer: 1216247-91-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR