4-Chloro-3-propoxyphenol 95%

Lieferant: Apollo Scientific

OR957573-1G
APOSOR957573-1GEA 632 EUR
APOSOR957573-1G
4-Chloro-3-propoxyphenol 95%
4-Chloro-3-propoxyphenol
Formel: C₉H₁₁ClO₂
Molecular Weight: 186,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440097
CAS-Nummer: 1378655-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR