N-Ethyl-4-hydroxybenzamide 95%

Lieferant: Apollo Scientific

OR957574-1G
APOSOR957574-1GEA 538 EUR
APOSOR957574-1G
N-Ethyl-4-hydroxybenzamide 95%
N-Ethyl-4-hydroxybenzamide
Formel: C₉H₁₁NO₂
Molecular Weight: 165,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11130754
CAS-Nummer: 27522-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR