4-Methoxy-2,3-dimethyl-phenol 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxy-2,3-dimethylphenol

OR957575-1G
APOSOR957575-1GEA 661 EUR
APOSOR957575-1G
4-Methoxy-2,3-dimethyl-phenol 95%
4-Methoxy-2,3-dimethyl-phenol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09032519
CAS-Nummer: 35355-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR