3-Hydroxy-2-methoxybenzoic acid 95%

Lieferant: Apollo Scientific

OR957576-1G
APOSOR957576-1GEA 632 EUR
APOSOR957576-1G
3-Hydroxy-2-methoxybenzoic acid 95%
3-Hydroxy-2-methoxybenzoic acid
Formel: C₈H₈O₄
Molecular Weight: 168,147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03840538
CAS-Nummer: 2169-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR