3,5-Dichloro-4-methoxyphenol 97%

Lieferant: Apollo Scientific

OR957578-1G
APOSOR957578-1GEA 364 EUR
APOSOR957578-1G
3,5-Dichloro-4-methoxyphenol 97%
3,5-Dichloro-4-methoxyphenol
Formel: C₇H₆Cl₂O₂
Molecular Weight: 193,027 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12405414
CAS-Nummer: 56680-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR