2-Methoxyquinolin-3-ol 95%

Lieferant: Apollo Scientific

Synonyme: 2-MEthoxyquinolin-3-ol

OR957579-1G
APOSOR957579-1GEA 702 EUR
APOSOR957579-1G
2-Methoxyquinolin-3-ol 95%
2-Methoxyquinolin-3-ol
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18414678
CAS-Nummer: 172605-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR