3-(1-Pyrrolidinylmethyl)phenol 95%

Lieferant: Apollo Scientific

OR957585-1G
APOSOR957585-1GEA 632 EUR
APOSOR957585-1G
3-(1-Pyrrolidinylmethyl)phenol 95%
3-(1-Pyrrolidinylmethyl)phenol
Formel: C₁₁H₁₅NO
Molecular Weight: 177,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01309098
CAS-Nummer: 69383-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR