4-(4-Morpholinyl)phenol 95%

Lieferant: Apollo Scientific

Synonyme: 4-Morpholinophenol

OR957586-1G
APOSOR957586-1GEA 144 EUR
APOSOR957586-1G
4-(4-Morpholinyl)phenol 95%
4-(4-Morpholinyl)phenol
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,2157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066933
CAS-Nummer: 6291-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR