2-(Piperidin-1-yl)pyrimidin-5-ol 95%

Lieferant: Apollo Scientific

OR957587-1G
APOSOR957587-1GEA 632 EUR
APOSOR957587-1G
2-(Piperidin-1-yl)pyrimidin-5-ol 95%
2-(Piperidin-1-yl)pyrimidin-5-ol
Formel: C₉H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18074292
CAS-Nummer: 809236-47-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR