3-(Cyclopentyloxy)phenol 95%

Lieferant: Apollo Scientific

OR957588-1G
APOSOR957588-1GEA 632 EUR
APOSOR957588-1G
3-(Cyclopentyloxy)phenol 95%
3-(Cyclopentyloxy)phenol
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11181912
CAS-Nummer: 163419-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR