6-(Piperazin-1-yl)pyridin-3-ol 95%

Lieferant: Apollo Scientific

OR957591-1G
APOSOR957591-1GEA 632 EUR
APOSOR957591-1G
6-(Piperazin-1-yl)pyridin-3-ol 95%
6-(Piperazin-1-yl)pyridin-3-ol
Formel: C₉H₁₃N₃O
Molecular Weight: 179,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18259527
CAS-Nummer: 287114-37-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR