4-Methyl-1-naphthol 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methylnaphthalen-1-ol

OR957594-1G
APOSOR957594-1GEA 173 EUR
APOSOR957594-1G
4-Methyl-1-naphthol 95%
4-Methyl-1-naphthol
Formel: C₁₁H₁₀O
Molecular Weight: 158,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060344
CAS-Nummer: 10240-08-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR