4,5-Dichloro-2-methylphenol 98%

Lieferant: Apollo Scientific

OR957601-1G
APOSOR957601-1GEA 364 EUR
APOSOR957601-1G
4,5-Dichloro-2-methylphenol 98%
4,5-Dichloro-2-methylphenol
Formel: C₇H₆Cl₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18970307
CAS-Nummer: 52780-67-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR