Propyl-3-hydroxybenzoate 95%

Lieferant: Apollo Scientific

OR957605-1G
APOSOR957605-1GEA 632 EUR
APOSOR957605-1G
Propyl-3-hydroxybenzoate 95%
Propyl-3-hydroxybenzoate
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424456
CAS-Nummer: 38567-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR