2-(4-Hydroxy-2,6-dimethylphenoxy)acetonitrile 95%
Lieferant: Apollo Scientific
OR957617-1G
APOSOR957617-1GEA
632
EUR
APOSOR957617-1G
2-(4-Hydroxy-2,6-dimethylphenoxy)acetonitrile 95%
2-(4-Hydroxy-2,6-dimethylphenoxy)acetonitrile
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23111033 CAS-Nummer: 1394917-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...