2-[(Cyclopentylamino)methyl]phenol 95%

Lieferant: Apollo Scientific

OR957620-1G
APOSOR957620-1GEA 451 EUR
APOSOR957620-1G
2-[(Cyclopentylamino)methyl]phenol 95%
2-[(Cyclopentylamino)methyl]phenol
Formel: C₁₂H₁₇NO
Molecular Weight: 191,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11139824
CAS-Nummer: 827328-86-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR