4-(1-Piperidinylmethyl)-phenol 95%

Lieferant: Apollo Scientific

OR957622-1G
APOSOR957622-1GEA 197 EUR
APOSOR957622-1G
4-(1-Piperidinylmethyl)-phenol 95%
4-(1-Piperidinylmethyl)-phenol
Formel: C₁₂H₁₇NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18453499
CAS-Nummer: 73152-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR