3-(4-Morpholinylmethyl)phenol 95%

Lieferant: Apollo Scientific

OR957624-1G
APOSOR957624-1GEA 0 EUR
APOSOR957624-1G
3-(4-Morpholinylmethyl)phenol 95%
3-(4-Morpholinylmethyl)phenol
Formel: C₁₁H₁₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09836691
CAS-Nummer: 87476-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR