4-(1H-Pyrazol-1-yl)phenol 95%

Lieferant: Apollo Scientific

OR957627-1G
APOSOR957627-1GEA 538 EUR
APOSOR957627-1G
4-(1H-Pyrazol-1-yl)phenol 95%
4-(1H-Pyrazol-1-yl)phenol
Formel: C₉H₈N₂O
Molecular Weight: 160,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19982342
CAS-Nummer: 16209-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR