2-Chloro-4-(cyclopropylmethoxy)phenol 98%
Lieferant: Apollo Scientific
OR957628-1G
APOSOR957628-1GEA
632
EUR
APOSOR957628-1G
2-Chloro-4-(cyclopropylmethoxy)phenol 98%
2-Chloro-4-(cyclopropylmethoxy)phenol
Formel:
C₁₀H₁₁ClO₂ Molecular Weight: 198,646 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17000227 CAS-Nummer: 860616-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...