1-(4-Chloro-3-hydroxyphenyl)ethanone 98%

Lieferant: Apollo Scientific

OR957632-1G
APOSOR957632-1GEA 538 EUR
APOSOR957632-1G
1-(4-Chloro-3-hydroxyphenyl)ethanone 98%
1-(4-Chloro-3-hydroxyphenyl)ethanone
Formel: C₈H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00034670
CAS-Nummer: 61124-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR